missing translation for 'onlineSavingsMsg'
Learn More

Ryanodine Receptor 2 Antibody, Novus Biologicals™

Product Code. 18407051 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407051 25 μL 25µL
18216057 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18407051 Supplier Novus Biologicals Supplier No. NBP19009125ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

Ryanodine Receptor 2 Polyclonal specifically detects Ryanodine Receptor 2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ryanodine Receptor 2
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500 - 1:1000, Immunohistochemistry Free-Floating -Reported in scientific literature (PMID:30483051).
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias arrhythmogenic right ventricular dysplasia 2, ARVC2, ARVD2, cardiac muscle ryanodine receptor-calcium release channel, Cardiac muscle-type ryanodine receptor, cardiac-type ryanodine receptor, hRYR-2, islet-type ryanodine receptor, kidney-type ryanodine receptor, ryanodine receptor 2, ryanodine receptor 2 (cardiac), RyR2, RYR-2, type 2 ryanodine receptor, VTSIP
Gene Symbols RYR2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:QDEVRGDGEEGERKPLEAALPSEDLTDLKELTEESDLLSDIFGLDLKREGGQYKLIPHNPNAGLSDLMSNPVPMPEVQEKFQEQKAKEEEKEEKEETKSEPE
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6262
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mehr anzeigen Weniger anzeigen

For Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.