missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ryanodine Receptor 2 Antibody (C3-33), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80143AF647
This item is not returnable.
View return policy
Description
Ryanodine Receptor 2 Monoclonal specifically detects Ryanodine Receptor 2 in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Chicken, Fish, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Ryanodine Receptor 2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| RYR2 | |
| Canine cardiac ryanodine receptor (RyR2) | |
| 0.1 mL | |
| Primary | |
| Detects Ryanodine Receptor 2. Also shows cross-reactivity with Ryanodine Receptor 1 in amphibian, canine, chicken, fish, guinea pig and rat tissues. | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| C3-33 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| arrhythmogenic right ventricular dysplasia 2, ARVC2, ARVD2, cardiac muscle ryanodine receptor-calcium release channel, Cardiac muscle-type ryanodine receptor, cardiac-type ryanodine receptor, hRYR-2, islet-type ryanodine receptor, kidney-type ryanodine receptor, ryanodine receptor 2, ryanodine receptor 2 (cardiac), RyR2, RYR-2, type 2 ryanodine receptor, VTSIP | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6262 | |
| Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Chicken, Fish, Guinea Pig, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction