missing translation for 'onlineSavingsMsg'
Få mere at vide

Emma Safety Footwear Ryan S3 Shoe

Artikelnummer. 17592526
Skift visning
Klik for at se tilgængelige muligheder
Size:
39
40
41
42
43
44
45
46
47
48
Pakningsstørrelse:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Size Pakningsstørrelse
17592526 43 1 pair
17552526 39 1 pair
17562526 40 1 pair
17572526 41 1 pair
17582526 42 1 pair
17502536 44 1 pair
17512536 45 1 pair
17522536 46 1 pair
17532536 47 1 pair
17542536 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17592526 Leverandør Emma Safety Footwear Leverandørnr. MM73556843
Stock Up and Save Up to 60%
Use promo code 28009
Se alle kampagner
Estimated Shipment: 02-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

The Ryan has everything you would expect from a safety boot; rugged and durable with all day long comfort and protection for your feet in all conditions.

  • Waterproof
  • The all-round safety boot: extremely safe & comfortable, 100% metal free & waterproof.
  • The Ryan has everything you would expect from a safety boot; rugged and durable with all day long comfort and protection for your feet in all conditions.
  • This boot has a fully waterproof membrane, so you don't have to worry about wet feet. You can fully concentrate on getting the job done.
  • The reflective parts on the boot ensure that you are clearly visible in the dark and the Gripforce outsole gives you maximum grip.
  • The extremely cool carbon look on the side of the upper gives the boot a sporty look.
  • It is available in 2 widths (D & XD).
  • Industries
    • Chemical industry
    • Metal industry
    • Construction & technology
    • Manufacturing
    • Nature & environment
  • Safety shoe
    • S3-Safetyshoe with sole plate for wet work

Tekniske data

Closure Type Laces
Color Black
Liner Material Hydro-Tec Antibacterial/DryShield Membrane
Material Leather
Size 43
Certifications/Compliance (Europe) ISO 20345:2011
Fit Extra-wide Width
Waterproof Yes
EN ISO 20345 S3
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.