missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RXRB Monoclonal Antibody (147)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA14669
This item is not returnable.
View return policy
Description
This antibody is specific for the ~48 kDa RXR-beta protein.
This gene encodes a member of the retinoid X receptor (RXR) family of nuclear receptors which are involved in mediating the effects of retinoic acid (RA). This receptor forms homodimers with the retinoic acid, thyroid hormone, and vitamin D receptors, increasing both DNA binding and transcriptional function on their respective response elements. The gene lies within the major histocompatibility complex (MHC) class II region on chromosome 6.
Specifications
| RXRB | |
| Monoclonal | |
| 0.5 mg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P28704, P49743 | |
| Rxrb | |
| Synthetic peptide corresponding to the hinge region of mouse RXRB conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 147 | |
| Unconjugated | |
| Rxrb | |
| AL023085; DADB-100D22.1; DAUDI6; H-2RIIBP; MGC1831; MHC class I promoter binding protein; MHC class I regulatory element-binding protein H-2RIIBP; NR2B2; nuclear receptor coregulator 1; nuclear receptor co-regulator 1; nuclear receptor subfamily 2 group B member 2; putative retinoid X receptor beta; RCoR-1; Retinoic acid receptor RXR-beta; retinoic acid receptor, beta; retinoid X receptor; retinoid X receptor beta; retinoid X receptor, beta; Rub; RXR Beta; Rxrb; RXR-beta | |
| Mouse | |
| Protein G | |
| RUO | |
| 20182, 361801 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction