missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXR gamma/NR2B3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98864CL1
This item is not returnable.
View return policy
Description
RXR gamma/NR2B3 Polyclonal antibody specifically detects RXR gamma/NR2B3 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| RXR gamma/NR2B3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| NR2B3retinoid X receptor-gamma, Nuclear receptor subfamily 2 group B member 3, retinoic acid receptor RXR-gamma, Retinoid X receptor gamma, retinoid X receptor, gamma, RXRC | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RXR gamma/NR2B3. | |
| 0.1 mL | |
| Cancer, Transcription Factors and Regulators | |
| 6258 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction