missing translation for 'onlineSavingsMsg'
Learn More

RXR gamma/NR2B3 Antibody, Biotin, Novus Biologicals™

Product Code. 18330333 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330333 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18330333 Supplier Novus Biologicals Supplier No. NBP298864B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RXR gamma/NR2B3 Polyclonal specifically detects RXR gamma/NR2B3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXR gamma/NR2B3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias NR2B3retinoid X receptor-gamma, Nuclear receptor subfamily 2 group B member 3, retinoic acid receptor RXR-gamma, Retinoid X receptor gamma, retinoid X receptor, gamma, RXRC
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human RXR gamma/NR2B3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6258
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.