missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXR beta/NR2B2 Mouse anti-Human, Clone: PCRP-RXRB-2B6, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07586-100ug
This item is not returnable.
View return policy
Description
RXR beta/NR2B2 Monoclonal specifically detects RXR beta/NR2B2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| RXR beta/NR2B2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Protein Array | |
| DAUDI6, H-2RIIBP, MHC class I promoter binding protein, NR2B2MGC1831, Nuclear receptor subfamily 2 group B member 2, RCoR-1, retinoic acid receptor RXR-beta, Retinoid X receptor beta, retinoid X receptor, beta | |
| Recombinant full-length human RXR beta/NR2B2 protein | |
| 100 μg | |
| Cancer, Neuroscience, Transcription Factors and Regulators, Vision | |
| 6257 | |
| Store at 4°C. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PCRP-RXRB-2B6 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction