missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXR beta/NR2B2 Antibody (PCRP-RXRB-2B6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08445CL1
This item is not returnable.
View return policy
Description
RXR beta/NR2B2 Monoclonal antibody specifically detects RXR beta/NR2B2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Protein Array,Immunocytochemistry/Immunofluorescence
Specifications
| RXR beta/NR2B2 | |
| Monoclonal | |
| CoraFluor 1 | |
| DAUDI6, H-2RIIBP, MHC class I promoter binding protein, NR2B2MGC1831, Nuclear receptor subfamily 2 group B member 2, RCoR-1, retinoic acid receptor RXR-beta, Retinoid X receptor beta, retinoid X receptor, beta | |
| Recombinant full-length human RXR beta/NR2B2 protein (Uniprot: P28702) | |
| 0.1 mL | |
| Cancer, Neuroscience, Transcription Factors and Regulators, Vision | |
| 6257 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| PCRP-RXRB-2B6 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction