missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXR beta/NR2B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | RXR beta/NR2B2 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18214495
|
Novus Biologicals
NBP2-57755 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668658
|
Novus Biologicals
NBP2-57755-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RXR beta/NR2B2 Polyclonal specifically detects RXR beta/NR2B2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RXR beta/NR2B2 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Neuroscience, Transcription Factors and Regulators, Vision | |
| DAUDI6, H-2RIIBP, MHC class I promoter binding protein, NR2B2MGC1831, Nuclear receptor subfamily 2 group B member 2, RCoR-1, retinoic acid receptor RXR-beta, Retinoid X receptor beta, retinoid X receptor, beta | |
| RXRB | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 6257 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VAGGEQQTPEPEPGEAGRDGMGDSGRDSRSP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title