missing translation for 'onlineSavingsMsg'
Learn More

RXR alpha/NR2B1 Rabbit anti-Human, Mouse, Rat, Clone: 6Y10M1, Novus Biologicals™

Product Code. 18345332 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345332 20 μg 20µL
18339065 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18345332 Supplier Novus Biologicals Supplier No. NBP31566720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RXR alpha/NR2B1 Monoclonal antibody specifically detects RXR alpha/NR2B1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXR alpha/NR2B1
Applications ChIP Assay, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 6Y10M1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FLJ00280, FLJ00318, FLJ16020, MGC102720, NR2B1FLJ16733, Nuclear receptor subfamily 2 group B member 1, retinoic acid receptor RXR-alpha, Retinoid X receptor alpha, retinoid X receptor, alpha
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RXR alpha/NR2B1 (P19793).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6256
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.