missing translation for 'onlineSavingsMsg'
Learn More

RXR alpha/NR2B1 Antibody (4D6), Novus Biologicals™

Product Code. 18321229 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321229 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321229 Supplier Novus Biologicals Supplier No. H00006256M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RXR alpha/NR2B1 Monoclonal antibody specifically detects RXR alpha/NR2B1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXR alpha/NR2B1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4D6
Conjugate Unconjugated
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Alias FLJ00280, FLJ00318, FLJ16020, MGC102720, NR2B1FLJ16733, Nuclear receptor subfamily 2 group B member 1, retinoic acid receptor RXR-alpha, Retinoid X receptor alpha, retinoid X receptor, alpha
Host Species Mouse
Immunogen RXRA (AAH07925, 1 a.a. ~ 165 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLGSWPARTFHPGACVSRRPSAPWKHTASGKDSPDLRFSEHGVSQEFWAGGLVAVLEMTPSPSPWGTQEGPAGMCSLWVVGWCPCRGAGVRDLVLVHAGVWCKHVCAVQRDACGESRTPAPPRKGGAVTSVLCLFLIKTFPLFSYKFASCKQVHKDPPLVKSGFE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6256
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.