missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXFP4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20550-1-AP-20UL
This item is not returnable.
View return policy
Description
GPR100 is a member of the rhodopsin family of G protein-coupled receptors (GPRs) and acts as a high affinity receptor for INSL5. Also acts as receptor for RLN3/relaxin-3, as well as bradykinin and kallidin. Binding of the ligand inhibits cAMP accumulation.Specifications
| RXFP4 | |
| Polyclonal | |
| Unconjugated | |
| RXFP4 | |
| G protein coupled receptor 100, GPCR142, GPR100, Relaxin 3 receptor 2, RLN3 receptor 2, RLN3R2, RXFP4, RXFPR4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 339403 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8TDU9 | |
| RXFP4 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction