missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RXFP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5141162
This item is not returnable.
View return policy
Description
RXFP3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
RXFP3 (Relaxin Family Peptide Receptor 3) is a Protein Coding gene. Diseases associated with RXFP3 include Schizotypal Personality Disorder and Chromosome 5P13 Duplication Syndrome. Among its related pathways are Signaling by GPCR and GPCRs, Other. Gene Ontology (GO) annotations related to this gene include G protein-coupled receptor activity and G protein-coupled peptide receptor activity. An important paralog of this gene is GPR132.
Specifications
| RXFP3 | |
| Polyclonal | |
| Unconjugated | |
| RXFP3 | |
| BC053073; g protein-coupled receptor SALPR; G protein-coupled receptor SALPR homolog; GPCR135; G-protein coupled receptor GPCR135; G-protein coupled receptor SALPR; MGC141998; MGC142000; relaxin 3 receptor 1; relaxin family peptide receptor 3; relaxin/insulin like family peptide receptor 3; relaxin/insulin-like family peptide receptor 3; Relaxin-3 receptor 1; relaxin-3/INSL7 receptor 1; RGD1311550; RLN3 receptor 1; Rln3r1; Rxfp3; RXFPR3; SALPR; somatostatin- and angiotensin-like peptide receptor; somatostatin and angiotensin-like peptide receptor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 239336 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8BGE9 | |
| RXFP3 | |
| Synthetic peptide within Mouse Relaxin 3 receptor 1 aa 300-352. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction