Learn More
Invitrogen™ RXFP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595582
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human SHG-44 whole cell. Flow: U251 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The activity of this relaxin receptor is mediated by G proteins leading to stimulation of adenylate cyclase and an increase of cAMP and may also be a receptor for Leydig insulin-like peptide. Expressed in embryonic and adult gonads of males and females, as well in male gubernarculum and in brain. Not detected in kidney, spleen and heart.
Specifications
| RXFP2 | |
| Polyclonal | |
| Unconjugated | |
| RXFP2 | |
| G protein coupled receptor affecting testicular descent; G protein-coupled receptor 106; Gpcr; GPR106; G-protein coupled receptor 106; G-protein coupled receptor affecting testicular descent; GREAT; INSL3 receptor; INSL3R; leucine-rich repeat-containing G protein-coupled receptor 8; leucine-rich repeat-containing G-protein coupled receptor 8; Lgr8; LGR8.1; Relaxin family peptide receptor 2; relaxin receptor 2; relaxin/insulin like family peptide receptor 2; relaxin/insulin-like family peptide receptor 2; Rxfp2; RXFPR2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 122042 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q8WXD0 | |
| RXFP2 | |
| A synthetic peptide corresponding to a sequence of human GPCR LGR8 (MIVFLVFKHLFSLRLITMFFLLHFIVLINVKDFALTQ). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.