missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RX 821002 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | RX 821002 Hydrochloride |
| CAS | 109544-45-8 |
| Molecular Formula | C12H14N2O3 • (HCl) |
| Quantity | 10 mg |
| Synonym | 2-Methoxyidazoxan Monohydrochloride, 1,4-Benzodioxin-1H-imidazole Deriv., 2-(2,3-Dihydro-2-methoxy-1,4-benzodioxin-2-yl)-4,5-dihydro-1H-imidazole Monohydrochloride |
| InChI Formula | InChI=1S/C12H14N2O3.ClH/c1-15-12(11-13-6-7-14-11)8-16-9-4-2-3-5-10(9)17-12;/h2-5H,6-8H2,1H3,(H,13,14);1H |
| SMILES | COC1(OC2=CC=CC=C2OC1)C3=NCCN3.Cl |
| IUPAC Name | 2-(2-methoxy-2,3-dihydrobenzo[b][1,4]dioxin-2-yl)-4,5-dihydro-1H-imidazole hydrochloride |
| Molecular Weight (g/mol) | 270.71 |
| Formula Weight | 270.08 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?