missing translation for 'onlineSavingsMsg'
Learn More

IKA™ RV 10.10 Model Glassware

Product Code. 11885942 Shop All IKA Products
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11885942 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11885942 Supplier IKA™ Supplier No. 0003755400
  • ONLINE EXCLUSIVE PRICE
    £1530.00
    £1493.00 / Pack of 1
    Save £37.00
    2% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Type Glassware
Includes 1000mL Evaporator and 1000mL Receiving flask
Quantity 1 Pack
Volume (Metric) 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.