missing translation for 'onlineSavingsMsg'
Learn More

Ruxolitinib Phosphate, TRC

Product Code. 30135821
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135821 100 mg 100mg
17925045 250 mg 250mg
17993345 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135821 Supplier Toronto Research Chemicals Supplier No. TRCR702005100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1092939-17-7
Molecular Formula C17 H18 N6 . H3 O4 P
Molecular Weight (g/mol) 404.36
Synonym 1H-Pyrazole-1-propanenitrile, β-cyclopentyl-4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-, (βR)-, phosphate (1:1), INC 424 phosphate, INCB 018424 phosphate, Jakafi, Ruxolitinib phosphate
IUPAC Name (3R)-3-cyclopentyl-3-[4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)pyrazol-1-yl]propanenitrile;phosphoric acid
SMILES OP(=O)(O)O.N#CC[C@H](C1CCCC1)n2cc(cn2)c3ncnc4[nH]ccc34

Tekniske data

Quantity 100 mg
InChI Formula InChI=1S/C17H18N6.H3O4P/c18-7-5-15(12-3-1-2-4-12)23-10-13(9-22-23)16-14-6-8-19-17(14)21-11-20-16;1-5(2,3)4/h6,8-12,15H,1-5H2,(H,19,20,21);(H3,1,2,3,4)/t15-;/m1./s1
Formula Weight 404.1362
Percent Purity >95%
Chemical Name or Material Ruxolitinib Phosphate
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.