missing translation for 'onlineSavingsMsg'
Learn More

Ruxolitinib phosphate, MedChemExpress

Product Code. p-200096204
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481939

Brand: Medchem Express HY50858

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ruxolitinib phosphate (INCB018424 phosphate) is a potent JAK1/2 inhibitor with IC50s of 3.3 nM/2.8 nM, respectively, showing more than 130-fold selectivity over JAK3.

Ruxolitinib phosphate (INCB018424 phosphate) is a potent JAK1/2 inhibitor with IC50s of 3.3 nM/2.8 nM, respectively, showing more than 130-fold selectivity over JAK3.

Specifications

Chemical Name or Material Ruxolitinib phosphate
Physical Form Solid
CAS 1092939-17-7
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C17H21N6O4P
For Use With (Application) Cancer-Kinase/protease
Synonym INCB018424 phosphate
Solubility Information DMSO : ≥ 31 mg/mL (76.66 mM) ∣H2O : 10 mg/mL (24.73 mM; Need ultrasonic)
SMILES N#CC[C@H](C1CCCC1)N2N=CC(C3=C4C=CNC4=NC=N3)=C2.O=P(O)(O)O
Molecular Weight (g/mol) 404.36
Formula Weight 404.36
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.