missing translation for 'onlineSavingsMsg'
Learn More

Ruxolitinib phosphate, MedChemExpress

Product Code. 16467876
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467876 100 mg 100mg
16481939 1 g 1g
16451926 500 mg 500mg
16441326 200 mg 200mg
16428118 10 mg 10mg
16420578 5 mg 5mg
16404037 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16467876 Supplier Medchem Express Supplier No. HY50858
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ruxolitinib phosphate (INCB018424 phosphate) is a potent JAK1/2 inhibitor with IC50s of 3.3 nM/2.8 nM, respectively, showing more than 130-fold selectivity over JAK3.

Ruxolitinib phosphate (INCB018424 phosphate) is a potent JAK1/2 inhibitor with IC50s of 3.3 nM/2.8 nM, respectively, showing more than 130-fold selectivity over JAK3.

Specifications

Chemical Name or Material Ruxolitinib phosphate
Physical Form Solid
CAS 1092939-17-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H21N6O4P
For Use With (Application) Cancer-Kinase/protease
Synonym INCB018424 phosphate
Solubility Information DMSO : ≥ 31 mg/mL (76.66 mM) ∣H2O : 10 mg/mL (24.73 mM; Need ultrasonic)
SMILES N#CC[C@H](C1CCCC1)N2N=CC(C3=C4C=CNC4=NC=N3)=C2.O=P(O)(O)O
Molecular Weight (g/mol) 404.36
Formula Weight 404.36
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.