missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUVBL2 Antibody (SR1845), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21767-100ul
This item is not returnable.
View return policy
Description
RUVBL2 Monoclonal antibody specifically detects RUVBL2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| RUVBL2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| SR1845 | |
| Western Blot 1:500-1:2000 | |
| 48 kDa TATA box-binding protein-interacting protein, 51 kDa erythrocyte cytosolic protein, EC 3.6.1, EC 3.6.4.12,48 kDa TBP-interacting protein, ECP51TIP49B, erythrocyte cytosolic protein, 51-KD, INO80 complex subunit J, INO80JTAP54-beta, Repressing pontin 52, REPTIN, Reptin 52, Reptin52, RuvB (E coli homolog)-like 2, ruvB-like 2, RuvB-like 2 (E. coli), RVB2, TBP-interacting protein, 48-KD, TIH2, TIP48ECP-51, TIP49b, TIP60-associated protein 54-beta | |
| A synthesized peptide derived from human RUVBL2 (Uniprot #: Q9Y230) | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10856 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction