missing translation for 'onlineSavingsMsg'
Learn More

RUVBL2 Antibody (OTI2B9), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18423036 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423036 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423036 Supplier Novus Biologicals Supplier No. NBP273960MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RUVBL2 Monoclonal antibody specifically detects RUVBL2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUVBL2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2B9
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias 48 kDa TATA box-binding protein-interacting protein, 51 kDa erythrocyte cytosolic protein, EC 3.6.1, EC 3.6.4.12,48 kDa TBP-interacting protein, ECP51TIP49B, erythrocyte cytosolic protein, 51-KD, INO80 complex subunit J, INO80JTAP54-beta, Repressing pontin 52, REPTIN, Reptin 52, Reptin52, RuvB (E coli homolog)-like 2, ruvB-like 2, RuvB-like 2 (E. coli), RVB2, TBP-interacting protein, 48-KD, TIH2, TIP48ECP-51, TIP49b, TIP60-associated protein 54-beta
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 113-370 of human RUVBL2(NP_006657) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10856
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.