missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RUVBL2 Polyclonal antibody specifically detects RUVBL2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | RUVBL2 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 48 kDa TATA box-binding protein-interacting protein, 51 kDa erythrocyte cytosolic protein, EC 3.6.1, EC 3.6.4.12,48 kDa TBP-interacting protein, ECP51TIP49B, erythrocyte cytosolic protein, 51-KD, INO80 complex subunit J, INO80JTAP54-beta, Repressing pontin 52, REPTIN, Reptin 52, Reptin52, RuvB (E coli homolog)-like 2, ruvB-like 2, RuvB-like 2 (E. coli), RVB2, TBP-interacting protein, 48-KD, TIH2, TIP48ECP-51, TIP49b, TIP60-associated protein 54-beta |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 1 and 50 of human RuvB-like 2 using the numbering given in entry NP_006657.1 (GeneID 10856). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?