missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUVBL1 Antibody (01), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06522-50ul
This item is not returnable.
View return policy
Description
RUVBL1 Monoclonal antibody specifically detects RUVBL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RUVBL1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 01 | |
| Immunohistochemistry-Paraffin 1:20000-1:100000 | |
| 49 kDa TATA box-binding protein-interacting protein, 54 kDa erythrocyte cytosolic protein, EC 3.6.1, EC 3.6.4.12,49 kDa TBP-interacting protein, ECP54, INO80 complex subunit H, INO80HTIP49A, NMP 238, NMP238ECP-54, Nuclear matrix protein 238, PONTIN, Pontin 52, Pontin52, RuvB (E coli homolog)-like 1, ruvB-like 1, RuvB-like 1 (E. coli), RVB1, TATA binding protein interacting protein 49 kDa, TIH1, TIP49a, TIP49TAP54-alpha, TIP60-associated protein 54-alpha | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RUVBL1 (Uniprot#: Q9Y265-1; Met1-Lys456). | |
| 50 μg | |
| Core ESC Like Genes, Stem Cell Markers, Wnt Signaling Pathway | |
| 8607 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction