missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ruv B Antibody (RuvB 4G5/1), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50279AF532
This item is not returnable.
View return policy
Description
Ruv B Monoclonal antibody specifically detects Ruv B in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Ruv B | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| DNA Repair, Epigenetics, Virology Bacteria and Parasites | |
| E. coli | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry | |
| RuvB 4G5/1 | |
| Western Blot, ELISA, Immunohistochemistry | |
| EC 3.1.22.4, Holliday junction branch migration complex subunit RuvB | |
| Ruv B protein from E. Coli | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction