missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rutin, MedChemExpress
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Description
Rutin (Rutoside) is a flavonoid found in many plants and shows a wide range of biological activities including anti-inflammatory, antidiabetic, antioxidant, neuroprotective, nephroprotective, hepatoprotective and reducing Aβ oligomer activities. Rutin can cross the blood brain barrier. Rutin attenuates vancomycin-induced renal tubular cell apoptosis via suppression of apoptosis, mitochondrial dysfunction, and oxidative stress.
Specifications
Specifications
| Chemical Name or Material | Rutin |
| CAS | 153-18-4 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C27H30O16 |
| Quantity | 10 g |
| Synonym | Rutoside Quercetin 3-O-rutinoside |
| Solubility Information | DMSO : 50 mg/mL (81.90 mM; Need ultrasonic) |
| SMILES | O=C1C(O[C@H]2[C@@H]([C@H]([C@@H]([C@@H](CO[C@H]3[C@@H]([C@@H]([C@H]([C@H](C)O3)O)O)O)O2)O)O)O)=C(C4=CC=C(O)C(O)=C4)OC5=CC(O)=CC(O)=C15 |
| Molecular Weight (g/mol) | 610.52 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction