missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium standard solution, for AAS, 1mg/mL Ru in 10-20% HCl, Thermo Scientific Chemicals

Product Code. 10266440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10266440 100 mL 100mL
11964081 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10266440 Supplier Thermo Scientific Acros Supplier No. 196251000
  • ONLINE EXCLUSIVE PRICE
    £33.10
    £30.90 / 100mL
    Save £2.20
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-18-8 | Molecular Formula: Ru | Molecular Weight: 101.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ruthenium standard solution
Name Note 1 mg/ml Ru in 5% HCl
Packaging Plastic bottle
CAS 7440-18-8, 7647-01-0, 7732-18-5
Density 1.0200g/mL
Specific Gravity 1.02
Physical Form Solution
Molecular Formula Ru
Concentration 990 to 1010 μg/mL of Ru(3+) in 10−20% HCl
MDL Number MFCD00011207 MFCD03458417
Color Yellow to Brown
Melting Point -25.0°C
Boiling Point 107.0°C
Solubility Soluble in water
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES [Ru]
IUPAC Name ruthenium
Molecular Weight (g/mol) 101.07
PubChem CID 23950
Quantity 100 mL
ChEBI CHEBI:30682
Formula Weight 101.07
Grade AAS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ruthenium atomic absorption standard solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.