Learn More
Ruthenium powder, -22 mesh, Premion, 99.98% (metals basis excluding Ca), Thermo Scientific Chemicals
£520.00 - £2289.00
Chemical Identifiers
CAS | 7440-18-8 |
---|---|
Molecular Formula | Ru |
Molecular Weight (g/mol) | 101.07 |
MDL Number | MFCD00011207 MFCD03458417 |
InChI Key | KJTLSVCANCCWHF-UHFFFAOYSA-N |
Synonym | on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon |
PubChem CID | 23950 |
ChEBI | CHEBI:30682 |
IUPAC Name | ruthenium |
SMILES | [Ru] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11310899
|
Thermo Scientific Alfa Aesar
010667.06 |
5 g |
£520.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11390889
|
Thermo Scientific Alfa Aesar
010667.14 |
25 g |
£2289.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-18-8 | |
101.07 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
23950 | |
ruthenium |
Ru | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
CHEBI:30682 | |
[Ru] |
Specifications
7440-18-8 | |
Ru | |
UN3089 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium | |
23950 | |
99.98% | |
Odorless | |
Ruthenium powder |
Powder | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
[Ru] | |
101.07 | |
CHEBI:30682 | |
Premion™ | |
(metals basis excluding Ca), Ca 100ppm max. |
Safety and Handling
GHS H Statement
H228
Flammable solid.
GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only