missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11384758
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384758 1 g 1g
11394758 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384758 Supplier Thermo Scientific Alfa Aesar Supplier No. 043098.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ruthenium(IV) sulfide is used to prepare sintered material, which is a semiconductor. It acts as an electrode and posses high catalytic activity for oxygen evolution from aqueous solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12166-20-0
Molecular Formula RuS2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00799846
InChI Key XNQLQKWNBPVVGB-UHFFFAOYSA-N
Synonym ruthenium iv sulfide, ruthenium sulfide, bis sulfanylidene ruthenium, dithioxoruthenium, ruthenium disulphide, disulfanylideneruthenium
PubChem CID 82958
SMILES S=[Ru]=S

Specifications

Physical Form Powder
Quantity 1 g
UN Number UN3178
IUPAC Name disulfanylideneruthenium
Formula Weight 165.19
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Ruthenium (IV) sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.