missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) Oxide Hydrate, TRC

Product Code. 30423414
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423414 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423414 Supplier Toronto Research Chemicals Supplier No. TRCR70181510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ruthenium(IV) Oxide Hydrate
CAS 32740-79-7
Quantity 10 g
Molecular Formula O2Ru . X(H2O)
Synonym Hydrous Ruthenium Oxide, Ruthenium Dioxide Hydrate
InChI Formula InChI=1S/CH2O.2H2O.Ru/c1-2;;;/h1H2;2*1H2;
SMILES O.O.[Ru].C=O
IUPAC Name formaldehyde;ruthenium;dihydrate
Formula Weight 167.936
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.