missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) oxide hydrate, Ru 54% min

Product Code. 11351838
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11351838

Brand: Thermo Scientific Alfa Aesar 011803.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32740-79-7 | O2Ru | 133.07 g/mol

OxidationRuthenium(IV) oxide hydrate is used as a starting material for the preparation of ruthenium tetraoxide by oxidation. It is also used for the coating of titanium anodes and for the preparation of resistors. It is an active material in supercapacitor due to its high charge transfer capability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32740-79-7
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00149846
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O

Specifications

Melting Point 1200° (sublimation)
Color Black
Physical Form Powder
Quantity 5 g
Assay Percent Range Ru 54% min.
Solubility Information Insoluble in water. Soluble in HClInsoluble in water.
Formula Weight 133.07 (Anhydrous)
Odor Odorless
Appearance Black powder
Chemical Name or Material Ruthenium(IV) oxide hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.