missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals

Catalog Number 11335687
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
32740-79-7
O2Ru
133.07
MFCD00149846
WOCIAKWEIIZHES-UHFFFAOYSA-N
dioxoruthenium
O=[Ru]=O
This item is not returnable. View return policy
32740-79-7
O2Ru
133.07
MFCD00149846
WOCIAKWEIIZHES-UHFFFAOYSA-N
dioxoruthenium
O=[Ru]=O

Chemical catalyst, important material for producing resistance and capacitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1200°C (sublimation)
Color Black
Physical Form Powder
Quantity 1 g
UN Number UN1479
Solubility Information It is soluble in fused alkalis. It is insoluble in water.
Formula Weight 133.07 (Anhydrous)
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Ru 54 to 60%
Assay (metals basis)
Chemical Name or Material Ruthenium (IV) oxide hydrate
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'