Learn More
Ruthenium(IV) oxide, Electronic Grade, Premion™, 99.95% (metals basis), Ru typically 74%, Thermo Scientific Chemicals
For Semiconductor Industry Use For the Manufacture of Resistor Pastes
£156.00 - £779.00
Chemical Identifiers
CAS | 12036-10-1 |
---|---|
Molecular Formula | O2Ru |
Molecular Weight (g/mol) | 133.07 |
MDL Number | MFCD00011210 |
InChI Key | WOCIAKWEIIZHES-UHFFFAOYSA-N |
Synonym | ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous |
PubChem CID | 82848 |
IUPAC Name | dioxoruthenium |
SMILES | O=[Ru]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313908
|
Thermo Scientific Alfa Aesar
040336.04 |
2 g |
£156.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393898
|
Thermo Scientific Alfa Aesar
040336.09 |
10 g |
£779.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. It is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12036-10-1 | |
133.07 | |
WOCIAKWEIIZHES-UHFFFAOYSA-N | |
82848 | |
O=[Ru]=O |
O2Ru | |
MFCD00011210 | |
ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous | |
dioxoruthenium |
Specifications
1200° (sublimation) | |
Black | |
99.95% (metals basis) | |
MFCD00011210 | |
Insoluble in water,acids. Soluble in fused alkalis. | |
O=[Ru]=O | |
133.07 | |
133.07 | |
Odorless | |
Black powder | |
Ruthenium(IV) oxide |
12036-10-1 | |
Sub-micron Powder | |
O2Ru | |
ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous | |
WOCIAKWEIIZHES-UHFFFAOYSA-N | |
dioxoruthenium | |
82848 | |
Electronic | |
Hygroscopic | |
6.97 |
Safety and Handling
P264b-P280i-P305+P351+P338
H319
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II
missing translation for 'einecsNumber' : 234-840-6
missing translation for 'rtecsNumber' : VM2654000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only