missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) Oxide Anhydrous, TRC

Product Code. 30409925
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409925 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409925 Supplier Toronto Research Chemicals Supplier No. TRCR7018201G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ruthenium(IV) Oxide Anhydrous
CAS 12036-10-1
Quantity 1 g
Molecular Formula O2Ru
Synonym Ruthenium Oxide, NW 120, NW 47, NW 54, NW 67, Ruthenia, Ruthenium dioxide, Ruthenium(IV) dioxide, Ruthenium(IV) oxide, Ruthenium-Adams Catalyst
InChI Formula InChI=1S/2O.Ru
SMILES O=[Ru]=O
IUPAC Name dioxoruthenium
Molecular Weight (g/mol) 133.07
Formula Weight 133.8942
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.