missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Ru 75.2% min

Product Code. 11391838
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391838 2 g 2g
11381838 0.5 g 0.5g
11301848 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11391838 Supplier Thermo Scientific Alfa Aesar Supplier No. 011804.04
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 12036-10-1 | O2Ru | 133.07 g/mol

The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-10-1
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00011210
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
Synonym ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
PubChem CID 82848
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O

Specifications

Melting Point 1200° (sublimation)
Color Black
Physical Form Powder
Quantity 2 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Insoluble in water,and acids. Soluble in fused alkali.
Formula Weight 133.07
Odor Odorless
Sensitivity Hygroscopic
Appearance Black powder
Density 6.97 g/cm3 at 20°C
Chemical Name or Material Ruthenium(IV) oxide, anhydrous, Ru 75.2% min.
Show More Show Less
compliance-icons
Product Identifier
  • Ruthenium(IV) oxide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Ruthenium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.