missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v

Product Code. 11367477
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
50mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11367477

Brand: Thermo Scientific Alfa Aesar 012630.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 34513-98-9

Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34513-98-9
MDL Number MFCD00016313

Specifications

Color Red to Brown
Linear Formula Ru(NO)(NO3)3
UN Number UN3093
Concentration Ru 1.5% w/v
Density 1.07 g/mL
Solubility Information Miscible with cold water.
Formula Weight 317.09
Odor Acidic
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Ruthenium(III) nitrosylnitrate solution
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.