missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Description
Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Red to Brown |
Linear Formula | Ru(NO)(NO3)3 |
UN Number | UN3093 |
Concentration | Ru 1.5% w/v |
Density | 1.07 g/mL |
Solubility Information | Miscible with cold water. |
Formula Weight | 317.09 |
Odor | Acidic |
Quantity | 50 mL |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'