missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) nitrosylnitrate, Ru 31.3% min

Product Code. 11356558
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356558 25 g 25g
11366558 1 g 1g
11346558 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356558 Supplier Thermo Scientific Alfa Aesar Supplier No. 012175.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34513-98-9 | N4O10Ru

Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34513-98-9
Molecular Formula N4O10Ru
MDL Number MFCD00016313

Specifications

Melting Point (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 25 g
Assay Percent Range Ru 31.3% min.
Solubility Information Miscible with cold water.
Formula Weight 317.09
Odor Odorless
Appearance Red-brown crystals
Chemical Name or Material Ruthenium(III) nitrosylnitrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.