missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution

Product Code. 10228853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10228853 500 mL Glass bottle 500mL
10793403 25 mL Glass bottle 25mL
10089051 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10228853 Supplier Thermo Scientific Acros Supplier No. 365045000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34513-98-9 | N4O10Ru | 317.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34513-98-9
Molecular Formula N4O10Ru
Molecular Weight (g/mol) 317.09
MDL Number MFCD00016313

Specifications

Color Brown to Orange
Physical Form Solution
Quantity 500 mL
Solubility Information Solubility in water: soluble
Formula Weight 317.09
Percent Purity 1.5% (Ru) (typically)
Packaging Glass bottle
Specific Gravity 1.07
Density 1.0700g/mL
Chemical Name or Material Ruthenium(III) nitrosyl nitrate, In diluted nitric acid
compliance-icons
Product Identifier
  • Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.