Learn More
Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution, Thermo Scientific Chemicals
£61.70 - £971.00
Chemical Identifiers
CAS | 34513-98-9 |
---|---|
Molecular Formula | N4O10Ru |
Molecular Weight (g/mol) | 317.09 |
MDL Number | MFCD00016313 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10793403
|
Thermo Scientific Acros
365040250 |
25 mL | Glass bottle |
£61.70
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10089051
|
Thermo Scientific Acros
365041000 |
100 mL | Glass bottle |
£213.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10228853
|
Thermo Scientific Acros
365045000 |
500 mL | Glass bottle |
£971.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34513-98-9 | |
N4O10Ru | |
317.09 | |
MFCD00016313 |
Specifications
34513-98-9 | |
Solution | |
MFCD00016313 | |
317.09 | |
1.5% (Ru) (typically) | |
1.0700g/mL |
Brown to Orange | |
N4O10Ru | |
Solubility in water: soluble | |
317.09 | |
1.07 | |
Ruthenium(III) nitrosyl nitrate, In diluted nitric acid |
Sicherheit und Handhabung
GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
May intensify fire; oxidizer.
Corrosive to the respiratory tract.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 252-068-8
- Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution
Signalwort
- Danger
Gefahrenkategorie
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Gefahrenhinweis
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
Sicherheitshinweis
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only