missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) chloride hydrate, Premion™, 99.99% (metals basis)

Product Code. p-7058855
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11399538

Brand: Thermo Scientific Alfa Aesar 043364.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14898-67-0 | Cl3Ru | 207.42 g/mol

Ruthenium(III) chloride hydrate is used as a starting material in ruthenium chemistry. It acts as a precursor to many ruthenium compounds, which finds application as catalysts in asymmetric hydrogenation and alkene metathesis. It serves as a catalyst in the preparation of 2-ethyl-3-methylquinolines from primary aromatic amines and triallylamine. Further, it is used in the preparation of nanoparticulate ruthenium-aluminum oxyhydroxide catalyst for the efficient dehydrogenation of arylmethyl alcohols to the corresponding aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-67-0
Molecular Formula Cl3Ru
Molecular Weight (g/mol) 207.42
MDL Number MFCD00149844
InChI Key YBCAZPLXEGKKFM-UHFFFAOYSA-K
IUPAC Name ruthenium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Melting Point 100°C (decomposition)
Color Black
Physical Form Crystalline
Quantity 2 g
UN Number UN3260
Merck Index 14,8302
Solubility Information Soluble in water.
Formula Weight 207.43 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Ruthenium (III) chloride hydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ruthenium(III) chloride hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ruthenium(III) chlor

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.