missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) chloride hydrate, 99.9% (PGM basis), Ru 38% min

Product Code. 11381678
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381678 10 g 10g
11371678 2 g 2g
11391678 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11381678 Supplier Thermo Scientific Alfa Aesar Supplier No. 011043.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14898-67-0 | Cl3Ru | 207.42 g/mol

Oxidation.Ruthenium(III) chloride hydrate is used as a catalyst for alkyne functionalization and oxidation reactions. It is used as key raw materials for the preparation of various ruthenium-based catalysts, including Grubbs’ catalysts. It is utilized in the preparation of nanoparticulate ruthenium-aluminum oxyhydroxide catalyst, which is efficient for dehydrogenation of alcohols to the corresponding aldehydes. It is also involved in the synthesis of 2-ethyl-3-methylquinolines from primary aromatic amines and triallylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-67-0
Molecular Formula Cl3Ru
Molecular Weight (g/mol) 207.42
MDL Number MFCD00149844
InChI Key YBCAZPLXEGKKFM-UHFFFAOYSA-K
IUPAC Name ruthenium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Melting Point 100°dec
Physical Form Crystalline Soluble
Quantity 10 g
Assay Percent Range 99.9%
Solubility Information Very soluble in water. Soluble in alcohol,acetone.
Formula Weight 207.43 (Anhydrous)
Odor Pungent
Sensitivity Hygroscopic
Appearance Black crystalline or powder
Chemical Name or Material Ruthenium(III) chloride hydrate, PGM basis, Ru 38% min.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.