missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium black, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11351729
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
7440-18-8
Ru
101.07
MFCD00011207 MFCD03458417
KJTLSVCANCCWHF-UHFFFAOYSA-N
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
23950
CHEBI:30682
ruthenium
[Ru]
This item is not returnable. View return policy
7440-18-8
Ru
101.07
MFCD00011207 MFCD03458417
KJTLSVCANCCWHF-UHFFFAOYSA-N
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
23950
CHEBI:30682
ruthenium
[Ru]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 g
UN Number UN3089
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Ruthenium black

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'