missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RUSC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548330
This item is not returnable.
View return policy
Description
RUSC2 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a RUN and SH3 domain containing protein that interacts with Rab1b and Rab1-binding protein GM130. Alternatively spliced transcript variants have been observed for this gene.
Specifications
| RUSC2 | |
| Polyclonal | |
| Unconjugated | |
| RUSC2 | |
| AI840675; Interacting protein of Rab1; iporin; KIAA0375; mKIAA0375; RUN and SH3 domain containing 2; RUN and SH3 domain-containing protein 2; RUSC2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 9853 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8N2Y8 | |
| RUSC2 | |
| KLH conjugated synthetic peptide between 308-336 amino acids from the N-terminal region of human RUSC2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction