missing translation for 'onlineSavingsMsg'
Learn More

RUSC2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18318900 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18318900 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18318900 Supplier Novus Biologicals Supplier No. NBP281786

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RUSC2 Polyclonal specifically detects RUSC2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUSC2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias interacting protein of Rab1, Iporin, KIAA0375, RUN and SH3 domain containing 2, RUN and SH3 domain-containing protein 2
Gene Symbols RUSC2
Host Species Rabbit
Immunogen RUSC2 antibody was raised against an 18 amino acid synthetic peptide from near the amino terminus of human RUSC2. The immunogen is located within amino acids 70 - 120 of RUSC2. Amino Acid Squence: TSKRSRDGRGPGAPKRHN
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9853
Test Specificity At least three isoforms are known to exist; this antibody will detect all three isoforms. This antibody is predicted to not cross-react with RUSC1.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.