missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX3/CBFA3 Antibody (2B9G4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22390H
This item is not returnable.
View return policy
Description
RUNX3/CBFA3 Monoclonal specifically detects RUNX3/CBFA3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RUNX3/CBFA3 | |
| Monoclonal | |
| HRP | |
| NP_001026850.1 | |
| RUNX3 | |
| Amino acids 126-310 of human Runx3 were used as the immunogen for this antibody. | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 864 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2B9G4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Acute myeloid leukemia 2 protein, AML2SL3/AKV core-binding factor alpha C subunit, CBFA3MGC16070, CBF-alpha-3, PEA2 alpha C, PEA2-alpha C, PEBP2 alpha C, PEBP2A3FLJ34510, PEBP2-alpha C, runt domain, alpha subunit 3, runt-related transcription factor 3, SL3-3 enhancer factor 1 alpha C subunit, transcription factor AML2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction