missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX2 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 82636-2-RR-100UL
This item is not returnable.
View return policy
Description
The RUNX2 antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human RUNX2. This antibody recognizes human, mouse antigen. The RUNX2 antibody has been validated for the following applications: WB, IP, ELISA analysis.Specifications
| RUNX2 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| AML3; CBFA1; CCD; CCD1; MGC120022; MGC120023; OSF2; PEA2aA; PEBP2A1; PEBP2A2; PEBP2aA; PEBP2aA1 | |
| Peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| 57-60 kDa | |
| Liquid |
| Western Blot, Immunoprecipitation, ELISA | |
| 1G5 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 860 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction