Learn More
Invitrogen™ RUNX2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA141519
Product Code. 11524520
  • £462.00 / 100µg
Description
PA1-41519 detects RUNX2 in chicken, chimpanzee, new world monkey, equine, human and mouse samples. This antibody is predicted to react with rat, pufferfish, zebrafish, and xenopus based on sequence homology.
RUNX2, a nuclear protein, is a member of the RUNX family of transcription factors and has an Runt DNA-binding domain. It is essential for membranous and endochondral bone formation. It not only regulates osteoblastic differentiation and skeletal morphogenesis but also acts as a scaffold for nucleic acids and regulatory factors involved in skeletal gene expression. The protein can bind DNA both as a monomer and with more affinity, as a subunit of a heterodimeric complex. It plays a critical role in increasing TGFBR1 expression by osteoblasts and functions in cooperation with DLX5 or a related factor to activate osteoblast-specific gene expression. Mutations lead to disorders in bone development like cleidocranial dysplasia (CCD).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RUNX2
Polyclonal
Unconjugated
RUNX2
Acute myeloid leukemia 3 protein; AKV core binding factor; Aml3; AML-3; Cbf; Cbfa1; Cbfa-1; CBF-alpha-1; CCD; CCD1; CLCD; core binding factor alpha 1; core binding factor alpha 1 subunit; core-binding factor subunit alpha-1; core-binding factor, runt domain, alpha subunit 1; LOW QUALITY PROTEIN: runt-related transcription factor 2; LS3; Oncogene AML-3; Osf2; OSF-2; osteoblast-specific transcription factor 2; PEA2aA; PEA2-alpha A; PEBP2 alpha A; PEBP2A; Pebp2a1; PEBP2aA; PEBP2-alpha A; Pebpa2a; Polyomavirus enhancer-binding protein 2 alpha A subunit; runt domain, alpha subunit 1; runt related transcription factor 2; runt-related transcription factor 2; RUNX 2; RUNX family transcription factor 2; Runx2; RUNX-2; SL3/AKV core-binding factor alpha A subunit; SL3-3 enhancer factor 1 alpha A subunit; transcription factor RUNX2; transcription factor RUNX2/CBFA1
Rabbit
Protein G
RUO
100033890, 12393, 373919, 860
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1.0 mg/mL
PBS with 0.05% sodium azide
Q08775, Q13950, Q9XSB7
RUNX2
Synthetic peptide corresponding to somewhere between amino acids 250-300 of human RUNX2.
100 μg
Primary
Human, Mouse, Monkey, Horse, Chicken