missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX2/CBFA1 Rabbit anti-Human, Mouse, Clone: 7O7J4, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £425.00
Specifications
| Antigen | RUNX2/CBFA1 |
|---|---|
| Clone | 7O7J4 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18300626
|
Novus Biologicals
NBP3-15434-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18355595
|
Novus Biologicals
NBP3-15434-100UL |
100 μg |
£425.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RUNX2/CBFA1 Monoclonal antibody specifically detects RUNX2/CBFA1 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| RUNX2/CBFA1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Epigenetics, Growth and Development, Stem Cells, Transcription Factors and Regulators | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 860 | |
| IgG | |
| Affinity purified |
| 7O7J4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Acute myeloid leukemia 3 protein, CBFA1, CBF-alpha-1, CCD1, CCDAML3, CLCD, Core-binding factor subunit alpha-1, core-binding factor, runt domain, alpha subunit 1, MGC120023, ML3, oncogene AML-3, OSF2, OSF-2, osteoblast-specific transcription factor 2, PEA2aA, PEA2-alpha A, PEBP2A, PEBP2aA, PEBP2-alpha A, polyomavirus enhancer-binding protein 2 alpha A subunit, runt domain, alpha subunit 1, runt related transcription factor 2, runt-related transcription factor 2, RUNX2, SL3/AKV core-binding factor alpha A subunit, SL3-3 enhancer factor 1 alpha A subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 336-439 of human RUNX2/CBFA1 (Q13950). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title