missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RUNX1T1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540064
This item is not returnable.
View return policy
Description
RUNX1T1 Polyclonal Antibody for ChIP, ChIP-seq, ELISA
RUNX1T1 is a putative zinc finger transcription factor and oncoprotein. In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5'-region of the RUNX1 gene fused to the 3'-region of this gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. The protein encoded by this gene is a putative zinc finger transcription factor and oncoprotein. In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5'-region of the RUNX1 gene fused to the 3'-region of this gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. Several transcript variants encoding multiple isoforms have been found for this gene.
Specifications
| RUNX1T1 | |
| Polyclonal | |
| Unconjugated | |
| RUNX1T1 | |
| acute myelogenous leukemia 1 translocation 1 protein; acute myelogenous leukemia 1 translocation 1, cyclin-D related; AML1-MTG8; AML1T1; Cbfa2t1; CBFA2T1 identified gene homolog; CBFA2T1 isoform r1t1-11a61; CBFA2T1 isoform r1t1-11a62; CBFA2T1 isoform r1t1-11a63; CBFA2T1 isoform r1t1-11a64; CBFA2T1 isoform r1t1-11a65; CBFA2T1 isoform r1t1-7a47; CBFA2T1 isoform r1t1-7a48; CBFA2T1 isoform r1t1-7a49; CBFA2T1 isoform r1t1-7a50; CBFA2T1 isoform r1t1-7a51; CBFA2T1 isoform r1t1-7a52; CBFA2T1 isoform r1t1-7d53; CBFA2T1 isoform r1t1-7d54; CBFA2T1 isoform r1t1-7d55; CBFA2T1 isoform r1t1-7d56; CBFA2T1 isoform r1t1-8a57; CBFA2T1 isoform r1t1-8a58; CBFA2T1 isoform r1t1-8a59; CBFA2T1 isoform r1t1-8a60; Cbfa2t1h; CDR; core-binding factor, runt domain, alpha subunit 2, translocated to, 1; core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclin D-related; cyclin D-related; Cyclin-D-related protein; Eight twenty one protein; ETO; LOW QUALITY PROTEIN: protein CBFA2T1; MGC2796; MTG8; MTG8b; myeloid translocation gene on 8q22; protein CBFA2T1; protein CBFA2T1; LOW QUALITY PROTEIN: protein CBFA2T1; protein CBFA2T1-like; Protein ETO; Protein MTG8; runt related transcription factor 1; translocated to, 1 (cyclin D related); runt-related transcription factor 1; translocated to, 1 (cyclin D-related); RUNX1 partner transcriptional co-repressor 1; RUNX1 translocation partner 1; Runx1t1; si:ch211-232j17.1; translocated to, 1; wu:fi14b07; zgc:154044; zinc finger MYND domain-containing protein 2; ZMYND2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ChIP Assay, ChIP sequencing (ChIP-seq), ELISA | |
| Conc. Not Determined | |
| whole serum with 0.05% sodium azide | |
| Q06455 | |
| RUNX1T1 | |
| Two KLH-conjugated synthetic peptides containing sequences from the N-terminal and the central region of the protein, respectively corresponding to human ETO (runt-related transcription factor 1; translocated to, 1 (cyclin D-related). | |
| RUO | |
| 862 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction