Learn More
RUNX1T1 Mouse anti-Human, Unconjugated, Clone: 2G2F7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67086-1-IG-150UL
 View more versions of this product
Product Code. 16899505
  • £349.00 / 150µL
Description
The RUNX1T1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RUNX1T1. This antibody recognizes human, mouse, rat, pig antigen. The RUNX1T1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RUNX1T1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, ELISA
2G2F7
Unconjugated
150 μL
862
IgG1